Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
528833
Gene Name
NOP9 nucleolar protein
Gene Aliases
C10H14orf21
Chromosome Location
Chr.10: 20865333 - 20871269 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103265.1 NP_001096735.1 4 - 5 1044 90
GenBank mRNA BC140558.1 - 4 - 5 1044 90

View Full Product Details