Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGB3
Entrez Gene ID
613729
Gene Name
high mobility group box 3
Gene Aliases
-
Chromosome Location
Chr.X: 33603508 - 33611398 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113257.2 NP_001106728.1 1 - 2 77 65
GenBank mRNA EE334754.1 - 1 - 2 77 65

View Full Product Details