Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC4
Entrez Gene ID
781806
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
Chromosome Location
Chr.11: 72189744 - 72192917 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102324.1 NP_001095794.1 4 - 5 488 60
GenBank mRNA BC149374.1 - 4 - 5 488 60

View Full Product Details