Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNASE1L2
Entrez Gene ID
787784
Gene Name
deoxyribonuclease 1 like 2
Gene Aliases
-
Chromosome Location
Chr.25: 1761110 - 1764865 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105019.1 NP_001098489.1 5 - 6 520 64
GenBank mRNA BC142236.1 - 5 - 6 520 64

View Full Product Details