Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PPP1R1A
Entrez Gene ID
767949
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1A
Gene Aliases
-
Chromosome Location
Chr.5: 25506930 - 25514982 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130760.1 NP_001124232.1 2 - 3 254 82
GenBank mRNA BC108116.1 - 2 - 3 254 82

View Full Product Details