Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC85B
Entrez Gene ID
540332
Gene Name
coiled-coil domain containing 85B
Gene Aliases
-
Chromosome Location
Chr.29: 44005578 - 44006861 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144087.3 NP_001137559.1 1 - 1 1149 102

View Full Product Details