Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2E
Entrez Gene ID
539121
Gene Name
lysine methyltransferase 2E (inactive)
Gene Aliases
MLL5
Chromosome Location
Chr.4: 46202169 - 46293391 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082451.3 NP_001075920.2 5 - 6 872 84
GenBank mRNA BC133514.1 - 5 - 6 929 84

View Full Product Details