Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB24
Entrez Gene ID
539966
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
-
Chromosome Location
Chr.9: 40680757 - 40691831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205737.1 NP_001192666.1 2 - 3 1132 105

View Full Product Details