Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRD5A2
Entrez Gene ID
527024
Gene Name
steroid 5 alpha-reductase 2
Gene Aliases
-
Chromosome Location
Chr.11: 14322215 - 14370474 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205937.1 NP_001192866.1 2 - 3 446 96

View Full Product Details