Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP39
Entrez Gene ID
532363
Gene Name
Rho GTPase activating protein 39
Gene Aliases
-
Chromosome Location
Chr.14: 359335 - 416175 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205997.1 NP_001192926.1 8 - 9 2885 78

View Full Product Details