Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
517052
Gene Name
golgi membrane protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 79260862 - 79321427 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192463.1 NP_001179392.1 3 - 4 369 67

View Full Product Details