Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMB4
Entrez Gene ID
520448
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Gene Aliases
-
Chromosome Location
Chr.5: 43012654 - 43120774 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192523.1 NP_001179452.1 2 - 3 481 74

View Full Product Details