Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNT2
Entrez Gene ID
521089
Gene Name
cyclin T2
Gene Aliases
-
Chromosome Location
Chr.2: 62317936 - 62351985 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192531.1 NP_001179460.1 6 - 7 541 102

View Full Product Details