Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPAS1
Entrez Gene ID
528272
Gene Name
neuronal PAS domain protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 54078441 - 54094359 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192657.2 NP_001179586.2 7 - 8 953 56

View Full Product Details