Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSPRY
Entrez Gene ID
510681
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
Chromosome Location
Chr.8: 102605181 - 102629728 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206723.1 NP_001193652.1 3 - 4 533 75

View Full Product Details