Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZACN
Entrez Gene ID
789168
Gene Name
zinc activated ion channel
Gene Aliases
LGICZ1
Chromosome Location
Chr.19: 55578491 - 55582703 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191252.3 NP_001178181.1 8 - 9 1042 107

View Full Product Details