Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCNJ
Entrez Gene ID
523293
Gene Name
cyclin J
Gene Aliases
-
Chromosome Location
Chr.26: 17413418 - 17431419 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191472.1 NP_001178401.1 4 - 5 738 81

View Full Product Details