Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHLRC3
Entrez Gene ID
506625
Gene Name
NHL repeat containing 3
Gene Aliases
-
Chromosome Location
Chr.12: 23178047 - 23219179 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192100.1 NP_001179029.1 3 - 4 598 111

View Full Product Details