Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PJVK
Entrez Gene ID
618114
Gene Name
pejvakin
Gene Aliases
DFNB59
Chromosome Location
Chr.2: 18429578 - 18437394 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193183.1 NP_001180112.1 1 - 2 211 127

View Full Product Details