Product Details

SNP ID
rs75370563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123069622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTGACTTCTCACTGTATTCTC[A/G]TATGATAGAAAGAAGACTAGACAGT
Phenotype
MIM: 616942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDIA5 PubMed Links

Gene Details

Gene
PDIA5
Gene Name
protein disulfide isomerase family A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006810.3 Intron NP_006801.1

View Full Product Details