Product Details
- SNP ID
-
rs57875551
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:97946604 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGATGTATATATATATATATAT[A/C]TATCTATTCATGTATATTCATATAT
- Phenotype
-
MIM: 612049
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRYBG3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201163224] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRYBG3
- Gene Name
- crystallin beta-gamma domain containing 3
There are no transcripts associated with this gene.
- Gene
- MINA
- Gene Name
- MYC induced nuclear antigen
View Full Product Details