Product Details

SNP ID
rs76736373
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81974768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTGAGGACTAAAGGCGAGAGCT[A/G]TGTCTCCAAGCTTCCTTTGAGTTTT
Phenotype
MIM: 610800
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATG10 PubMed Links

Gene Details

Gene
ATG10
Gene Name
autophagy related 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131028.1 Intron NP_001124500.1
NM_031482.4 Intron NP_113670.1
XM_005248610.4 Intron XP_005248667.1
XM_005248611.4 Intron XP_005248668.1
XM_005248612.3 Intron XP_005248669.1
XM_011543660.2 Intron XP_011541962.1
XM_011543661.2 Intron XP_011541963.1
XM_017009944.1 Intron XP_016865433.1

View Full Product Details