Product Details

SNP ID
rs77778701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55160486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCAATTTTTTGCTGTTACATG[G/T]TCATAAGTTGGAATTTAGTCTTCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145734.2 Intron NP_001139206.2
NM_001170402.1 Intron NP_001163873.1
NM_152623.2 Intron NP_689836.2
XM_011543218.2 Intron XP_011541520.1
Gene
GPX8
Gene Name
glutathione peroxidase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008397.3 Intron NP_001008398.2
NM_001306197.1 Intron NP_001293126.1
NM_001306198.1 Intron NP_001293127.1
NM_001306201.1 Intron NP_001293130.1
XM_006714631.2 Intron XP_006714694.1

View Full Product Details