Product Details

SNP ID
rs74466886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89638688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGTACCTGCTTTAACGTTAGC[G/T]TCGCTGGGGGTAAGCGGGAAGCAGC
Phenotype
MIM: 610583
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD6 PubMed Links

Gene Details

Gene
ANKRD6
Gene Name
ankyrin repeat domain 6
There are no transcripts associated with this gene.

Gene
LOC101929057
Gene Name
uncharacterized LOC101929057
There are no transcripts associated with this gene.

Gene
LYRM2
Gene Name
LYR motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020466.4 68 Missense Mutation AAG,ACG K10T NP_065199.1
Gene
MDN1
Gene Name
midasin AAA ATPase 1
There are no transcripts associated with this gene.

View Full Product Details