Product Details
- SNP ID
-
rs76202881
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:32581112 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGTGACAAAATTTTCTGATGAA[A/G]TCACTGTTTTGCTACCAAGATTCAA
- Phenotype
-
MIM: 612927
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AVL9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142183918] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AVL9
- Gene Name
- AVL9 cell migration associated
- Gene
- DPY19L1P1
- Gene Name
- DPY19L1 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details