Product Details

SNP ID
rs76293865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4860080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTCATTGGTATTCACCTGTTG[A/C]AAGGAAAATTCAGTAGCCTGTATGT
Phenotype
MIM: 607436 MIM: 611491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAPOLB PubMed Links

Gene Details

Gene
PAPOLB
Gene Name
poly(A) polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020144.4 1915 Missense Mutation TTG,TTT L577F NP_064529.4
Gene
RADIL
Gene Name
Rap associating with DIL domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018059.4 1915 Intron NP_060529.4

View Full Product Details