Product Details

SNP ID
rs79934382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76203335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCTGATGTCAGAAAGACATGG[G/T]ATCAAATCTAGGTTATGTGACCTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986711 PubMed Links

Gene Details

Gene
LOC107986711
Gene Name
uncharacterized LOC107986711
There are no transcripts associated with this gene.

Gene
SRRM3
Gene Name
serine/arginine repetitive matrix 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110199.2 Intron NP_001103669.1
NM_001291831.1 Intron NP_001278760.1

View Full Product Details