Product Details

SNP ID
rs74990161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122994935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGAAAGTAAGAGGTAAACACAA[A/T]AAATAAGACGGCAAGGGACCTCAGG
Phenotype
MIM: 604867
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R16 PubMed Links

Gene Details

Gene
TAS2R16
Gene Name
taste 2 receptor member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016945.2 766 Missense Mutation ATT,TTT I234F NP_058641.1

View Full Product Details