Product Details

SNP ID
rs78396555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97777939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGACTTGCAAGTACATCAGAG[A/G]ACGTAATTTGTAAAATCTAGGGCAT
Phenotype
MIM: 613296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAPTM4B PubMed Links

Gene Details

Gene
LAPTM4B
Gene Name
lysosomal protein transmembrane 4 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018407.4 Intron NP_060877.3

View Full Product Details