Product Details

SNP ID
rs80267243
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143368583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGTATAGTTTTTTTCTCCGTTT[C/T]CCGAACTTCTCCCGCACGCTCAGCG
Phenotype
MIM: 601031 MIM: 606387
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RHPN1 PubMed Links

Gene Details

Gene
RHPN1
Gene Name
rhophilin Rho GTPase binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052924.2 Intron NP_443156.2
XM_005250772.3 Intron XP_005250829.1
XM_005250773.3 Intron XP_005250830.1
XM_006716502.3 Intron XP_006716565.1
XM_011516817.2 Intron XP_011515119.1
XM_011516818.1 Intron XP_011515120.1
XM_011516819.2 Intron XP_011515121.1
XM_011516820.2 Intron XP_011515122.1
XM_011516821.2 Intron XP_011515123.1
XM_011516822.2 Intron XP_011515124.1
XM_011516823.2 Intron XP_011515125.1
XM_017013013.1 Intron XP_016868502.1
XM_017013014.1 Intron XP_016868503.1
XM_017013015.1 Intron XP_016868504.1
XM_017013016.1 Intron XP_016868505.1
Gene
RHPN1-AS1
Gene Name
RHPN1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TOP1MT
Gene Name
topoisomerase (DNA) I, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details