Product Details

SNP ID
rs79146297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55742408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTTTACCATAATTTGTTTTA[A/C]TATGTTTTACCATATTATGTGTTTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM68 PubMed Links

Gene Details

Gene
TMEM68
Gene Name
transmembrane protein 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286657.1 Intron NP_001273586.1
NM_001286660.1 Intron NP_001273589.1
NM_001286661.1 Intron NP_001273590.1
NM_152417.2 Intron NP_689630.1
XM_005251150.1 Intron XP_005251207.1
XM_011517454.1 Intron XP_011515756.1
XM_011517455.2 Intron XP_011515757.1
XM_017013040.1 Intron XP_016868529.1

View Full Product Details