Product Details

SNP ID
rs75681427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34728331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGCTCATTTTGTGGATAAGAA[C/T]GAGAGATGAAATGACTTCCACAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM205A PubMed Links

Gene Details

Gene
FAM205A
Gene Name
family with sequence similarity 205 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141917.1 Intron NP_001135389.1

View Full Product Details