Product Details

SNP ID
rs74623670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92829321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGAGAGGGAGCCTATGCTGC[A/C]TTGCATACATCTCCTGAGCTCTAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD19P PubMed Links

Gene Details

Gene
ANKRD19P
Gene Name
ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF484
Gene Name
zinc finger protein 484
There are no transcripts associated with this gene.

View Full Product Details