Product Details

SNP ID
rs78793974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14887015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGAAGGATTTTTCAGCCATCCT[A/G]AGGCAATGGGGAGTTAGTTGGCAGG
Phenotype
MIM: 606503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUV39H2 PubMed Links

Gene Details

Gene
SUV39H2
Gene Name
suppressor of variegation 3-9 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193424.1 Intron NP_001180353.1
NM_001193425.1 Intron NP_001180354.1
NM_001193426.1 Intron NP_001180355.1
NM_001193427.1 Intron NP_001180356.1
NM_024670.3 Intron NP_078946.1
XM_006717503.3 Intron XP_006717566.1
XM_011519662.2 Intron XP_011517964.1
XM_017016637.1 Intron XP_016872126.1

View Full Product Details