Product Details
- SNP ID
-
rs77893331
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:20156276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCCTCCTCGGAATCTGAGAAGT[A/C]CGAAGGTTTCCCGGGACTGCTCGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DBX1
PubMed Links
Gene Details
- Gene
- DBX1
- Gene Name
- developing brain homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001029865.2 |
970 |
Missense Mutation |
GAC,TAC |
D324Y |
NP_001025036.2 |
View Full Product Details