Product Details

SNP ID
rs80074245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33705038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGGTAAATGTGGTCCTATGTA[A/C]TTGCAGAGTAAGAATTTAAAAAAAT
Phenotype
MIM: 107271
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf91 PubMed Links

Gene Details

Gene
C11orf91
Gene Name
chromosome 11 open reading frame 91
There are no transcripts associated with this gene.

Gene
CD59
Gene Name
CD59 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000611.5 5650 UTR 3 NP_000602.1
NM_001127223.1 5650 UTR 3 NP_001120695.1
NM_001127225.1 5650 UTR 3 NP_001120697.1
NM_001127226.1 5650 UTR 3 NP_001120698.1
NM_001127227.1 5650 UTR 3 NP_001120699.1
NM_203329.2 5650 UTR 3 NP_976074.1
NM_203330.2 5650 UTR 3 NP_976075.1
NM_203331.2 5650 UTR 3 NP_976076.1

View Full Product Details