Product Details

SNP ID
rs74372815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8603566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGACATCAACCCATTATTAGG[G/T]TGTCTTGTTCACAGTCCTCCTGCCC
Phenotype
MIM: 605257
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AICDA PubMed Links

Gene Details

Gene
AICDA
Gene Name
activation-induced cytidine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520772.2 1483 UTR 3 XP_011519074.1
XM_011520773.2 1483 Intron XP_011519075.1

View Full Product Details