Product Details

SNP ID
rs78875772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31384562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGTATCAACGGAATAAGTACA[C/T]GGGCTCACAAGGTGACTGAAGGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND5B PubMed Links

Gene Details

Gene
DENND5B
Gene Name
DENN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308339.1 7165 UTR 3 NP_001295268.1
NM_144973.3 7165 UTR 3 NP_659410.3
XM_005253326.2 7165 UTR 3 XP_005253383.1
XM_005253327.3 7165 UTR 3 XP_005253384.1
XM_005253330.3 7165 UTR 3 XP_005253387.1
XM_006719040.3 7165 UTR 3 XP_006719103.1
XM_011520590.2 7165 UTR 3 XP_011518892.1
XM_017018896.1 7165 UTR 3 XP_016874385.1
XM_017018897.1 7165 UTR 3 XP_016874386.1
XM_017018898.1 7165 UTR 3 XP_016874387.1
XM_017018899.1 7165 UTR 3 XP_016874388.1

View Full Product Details