Product Details

SNP ID
rs78845384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95580628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAACAATAACTACATCGCAAAG[C/T]TGTTGTAATTAAATGAATTAGTGCG
Phenotype
MIM: 608671
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 5065 Intron NP_001153572.1
NM_006984.4 5065 Intron NP_008915.1
NM_182848.3 5065 Intron NP_878268.1
XM_011521134.2 5065 Intron XP_011519436.1
XM_017020843.1 5065 Intron XP_016876332.1
XM_017020844.1 5065 Intron XP_016876333.1
Gene
DZIP1
Gene Name
DAZ interacting zinc finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014934.4 5065 UTR 3 NP_055749.1
NM_198968.3 5065 UTR 3 NP_945319.1
XM_005254032.1 5065 Intron XP_005254089.1
XM_006719916.1 5065 Intron XP_006719979.1
XM_006719917.2 5065 Intron XP_006719980.1
XM_006719918.1 5065 Intron XP_006719981.1
XM_006719920.1 5065 Intron XP_006719983.1
XM_017020444.1 5065 Intron XP_016875933.1
XM_017020445.1 5065 Intron XP_016875934.1
XM_017020446.1 5065 Intron XP_016875935.1
XM_017020447.1 5065 Intron XP_016875936.1

View Full Product Details