Product Details

SNP ID
rs74776319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19175869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCAGTTGAAAAGTTAATCTCA[A/G]TTCCTTTTGTAGTTATTTCTTAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYT17 PubMed Links

Gene Details

Gene
SYT17
Gene Name
synaptotagmin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308157.1 Intron NP_001295086.1
NM_016524.3 Intron NP_057608.2
XM_005255362.3 Intron XP_005255419.1
XM_011545865.1 Intron XP_011544167.1
XM_011545866.2 Intron XP_011544168.1
XM_011545868.1 Intron XP_011544170.1
XM_011545869.2 Intron XP_011544171.1
XM_011545870.2 Intron XP_011544172.1
XM_011545871.2 Intron XP_011544173.1
XM_011545872.2 Intron XP_011544174.1
XM_017023280.1 Intron XP_016878769.1
XM_017023281.1 Intron XP_016878770.1

View Full Product Details