Product Details

SNP ID
rs78927166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66558338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCGATTAATGAAGTGGTTATTTT[G/T]GCCTTTGCTTGTAAGTGTTCAGTTT
Phenotype
MIM: 616074 MIM: 607884 MIM: 188250
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CKLF PubMed Links

Gene Details

Gene
CKLF
Gene Name
chemokine-like factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040138.2 380 Missense Mutation TGG,TTG W76L NP_001035228.1
NM_016326.3 380 Intron NP_057410.1
NM_016951.3 380 Missense Mutation TGG,TTG W76L NP_058647.1
NM_181640.2 380 Intron NP_857591.1
NM_181641.2 380 Missense Mutation TGG,TTG W76L NP_857592.1
Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202509.1 380 Intron NP_001189438.1
NM_001204098.1 380 Intron NP_001191027.1
NM_001204099.1 380 Missense Mutation TGG,TTG W76L NP_001191028.1
Gene
CMTM1
Gene Name
CKLF like MARVEL transmembrane domain containing 1
There are no transcripts associated with this gene.

Gene
TK2
Gene Name
thymidine kinase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details