Product Details
- SNP ID
-
rs75183825
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71367139 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTATTGTCTCACTTTAAATTGG[A/G]GGAAACTGAGGCACGGTAGTGGTGC
- Phenotype
-
MIM: 114051
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALB2
PubMed Links
Gene Details
- Gene
- CALB2
- Gene Name
- calbindin 2
View Full Product Details