Product Details

SNP ID
rs79507134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6863518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCTGCATCTAGTCAGCCATCA[C/T]TGGCTCCTCTCTAAATTTGCATTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX12P2 PubMed Links

Gene Details

Gene
ALOX12P2
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details