Product Details

SNP ID
rs76429053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8475100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCCTCAGCTGCAGTGTGGCC[A/G]CTATCCCTTCTATGCACAGACCAGC
Phenotype
MIM: 160776 MIM: 607538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH10 PubMed Links

Gene Details

Gene
MYH10
Gene Name
myosin, heavy chain 10, non-muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256012.1 6882 UTR 3 NP_001242941.1
NM_001256095.1 6882 UTR 3 NP_001243024.1
NM_005964.3 6882 UTR 3 NP_005955.3
XM_011523875.2 6882 UTR 3 XP_011522177.1
XM_011523878.2 6882 UTR 3 XP_011522180.1
XM_011523879.2 6882 UTR 3 XP_011522181.1
XM_011523880.2 6882 UTR 3 XP_011522182.1
XM_017024677.1 6882 UTR 3 XP_016880166.1
XM_017024678.1 6882 UTR 3 XP_016880167.1
XM_017024679.1 6882 UTR 3 XP_016880168.1
XM_017024680.1 6882 UTR 3 XP_016880169.1
XM_017024681.1 6882 UTR 3 XP_016880170.1
XM_017024682.1 6882 UTR 3 XP_016880171.1
Gene
NDEL1
Gene Name
nudE neurodevelopment protein 1 like 1
There are no transcripts associated with this gene.

View Full Product Details