Product Details

SNP ID
rs77537820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9577305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAATCCTAAGGAAGGTTTCTAA[A/G]TGATGAGGATTTCAGACCAGGACAG
Phenotype
MIM: 609804 MIM: 604203
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP52 PubMed Links

Gene Details

Gene
CFAP52
Gene Name
cilia and flagella associated protein 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080556.1 Intron NP_001074025.1
NM_145054.4 Intron NP_659491.4
XM_017024227.1 Intron XP_016879716.1
Gene
STX8
Gene Name
syntaxin 8
There are no transcripts associated with this gene.

View Full Product Details