Product Details

SNP ID
rs78927136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60446229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCTACTACAGTAAGAATTGAG[G/T]TAACAGCAAAATACCTTTTTGTTTC
Phenotype
MIM: 605324
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APPBP2 PubMed Links

Gene Details

Gene
APPBP2
Gene Name
amyloid beta precursor protein binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282476.1 2631 UTR 3 NP_001269405.1
NM_006380.3 2631 UTR 3 NP_006371.2
XM_011524198.2 2631 UTR 3 XP_011522500.1
XM_011524199.2 2631 UTR 3 XP_011522501.1

View Full Product Details