Product Details

SNP ID
rs75400620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60600746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCGACGGGCACGGCGGGCGGG[A/G]GGCGGCACAGTTTGCCCGGGAGCAC
Phenotype
MIM: 605100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPM1D PubMed Links

Gene Details

Gene
PPM1D
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003620.3 564 Missense Mutation GAG,GGG E111G NP_003611.1

View Full Product Details