Product Details

SNP ID
rs74561531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43969791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATGGAGCTCCCAGGGGAGTGG[C/T]GACCACTGTCTCTATGGTTCGGTTG
Phenotype
MIM: 604086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF155 PubMed Links

Gene Details

Gene
ZNF155
Gene Name
zinc finger protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260486.1 Intron NP_001247415.1
NM_001260487.1 Intron NP_001247416.1
NM_001260488.1 Intron NP_001247417.1
NM_003445.3 Intron NP_003436.3
NM_198089.2 Intron NP_932355.2
XM_005259215.3 Intron XP_005259272.1
XM_011527278.2 Intron XP_011525580.1
XM_011527279.2 Intron XP_011525581.1
XM_017027248.1 Intron XP_016882737.1
Gene
ZNF221
Gene Name
zinc finger protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297588.1 Intron NP_001284517.1
NM_001297589.1 Intron NP_001284518.1
NM_013359.2 Intron NP_037491.2
XM_017027232.1 Intron XP_016882721.1
XM_017027233.1 Intron XP_016882722.1

View Full Product Details