Product Details

SNP ID
rs80291173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:54210876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTCTCTTGTCACAGTGCATGG[C/T]GTATAGGAAACAGTAATTGTTAAAT
Phenotype
MIM: 604898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PFDN4 PubMed Links

Gene Details

Gene
PFDN4
Gene Name
prefoldin subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002623.3 Intron NP_002614.2
XM_017027879.1 Intron XP_016883368.1

View Full Product Details