Product Details
- SNP ID
-
rs78851478
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:18389773 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCGCTTCATGGTGCTTATTCATA[A/C]CAGCCACGGTTATGACGGGTCGATT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DZANK1
PubMed Links
Gene Details
- Gene
- DZANK1
- Gene Name
- double zinc ribbon and ankyrin repeat domains 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001099407.1 |
2242 |
Missense Mutation |
GGT,GTT |
G649V |
NP_001092877.1 |
XM_005260742.3 |
2242 |
Missense Mutation |
GGT,GTT |
G668V |
XP_005260799.1 |
XM_005260743.3 |
2242 |
Missense Mutation |
GGT,GTT |
G482V |
XP_005260800.1 |
XM_006723584.2 |
2242 |
Missense Mutation |
GGT,GTT |
G593V |
XP_006723647.1 |
XM_011529268.2 |
2242 |
Missense Mutation |
GGT,GTT |
G683V |
XP_011527570.1 |
XM_011529271.2 |
2242 |
Missense Mutation |
GGT,GTT |
G674V |
XP_011527573.1 |
XM_011529272.2 |
2242 |
Missense Mutation |
GGT,GTT |
G674V |
XP_011527574.1 |
XM_011529275.2 |
2242 |
Missense Mutation |
GGT,GTT |
G482V |
XP_011527577.1 |
XM_011529277.2 |
2242 |
Missense Mutation |
GGT,GTT |
G336V |
XP_011527579.1 |
XM_017027909.1 |
2242 |
Missense Mutation |
GGT,GTT |
G677V |
XP_016883398.1 |
XM_017027910.1 |
2242 |
Missense Mutation |
GGT,GTT |
G674V |
XP_016883399.1 |
XM_017027911.1 |
2242 |
Missense Mutation |
GGT,GTT |
G674V |
XP_016883400.1 |
XM_017027912.1 |
2242 |
Missense Mutation |
GGT,GTT |
G672V |
XP_016883401.1 |
XM_017027913.1 |
2242 |
Missense Mutation |
GGT,GTT |
G667V |
XP_016883402.1 |
XM_017027914.1 |
2242 |
Missense Mutation |
GGT,GTT |
G655V |
XP_016883403.1 |
XM_017027915.1 |
2242 |
Missense Mutation |
GGT,GTT |
G655V |
XP_016883404.1 |
XM_017027916.1 |
2242 |
Missense Mutation |
GGT,GTT |
G605V |
XP_016883405.1 |
XM_017027917.1 |
2242 |
Missense Mutation |
GGT,GTT |
G541V |
XP_016883406.1 |
XM_017027918.1 |
2242 |
Missense Mutation |
GGT,GTT |
G541V |
XP_016883407.1 |
XM_017027919.1 |
2242 |
Missense Mutation |
GGT,GTT |
G541V |
XP_016883408.1 |
XM_017027920.1 |
2242 |
Missense Mutation |
GGT,GTT |
G482V |
XP_016883409.1 |
XM_017027921.1 |
2242 |
Missense Mutation |
GGT,GTT |
G410V |
XP_016883410.1 |
- Gene
- LINC00851
- Gene Name
- long intergenic non-protein coding RNA 851
There are no transcripts associated with this gene.
View Full Product Details